Mesh Stack and Hang Bins, Quantum Storage Systems
# de catalogue 10804-239
Fournisseur: QUANTUM STORAGE SYSTEMS
Spécifications
- Dimensions:Exterior: 10 3/4 × 5 1/2 × 5"
Interior: 10 1/4 × 5 × 4 3/4" - Cat. no.:10804-239
- Supplier No.:QMB530C
Spécifications
A propos de cet article
These wire mesh bins offer a nickel-chrome finish and a 3/8” x 3/8” grid pattern allowing complete visibility of the bin’s contents.
- Unique mesh pattern provides a dust and dirt free storage environment
- Rear hanger allows them to hang from any louvered wall panel or rail system
Strong design allows superior hanging capacity for part storage where hanging is vital. A rear hanger allows the bin to hang from any louvered panel or rail system and optional side hanger which allows bins to stack one on top of the other.
Open hopper design allows for easy picking while allowing visual access to stored content. Two optional label holders are available for all Mesh Bins. Chrome color.
Customers Who Bought This Also Bought