Thermoelectric Effects Apparatus
# de catalogue 470121-250
Fournisseur: United Scientific Supplies
Spécifications
- Description:Thermoelectric Effects Apparatus
- Cat. no.:470121-250
- Supplier No.:THEA01
Spécifications
A propos de cet article
Investigate the thermal characteristics of three electrical thermometry devices
Explore the features and behavior of a closed loop digital temperature control system
- Investigate the thermal characteristics of three electrical thermometry devices
- Explore the features and behavior of a closed loop digital temperature control system
The Thermoelectric Effects Apparatus consists of an oven containing a cooling fan, a massive metal block and electric heater supported on two thin struts, the three samples, and the control system sensor. A control unit provides the heating current and houses the digital temperature controller. The oven and the control unit are connected by three multi-pole cables.
Customers Who Bought This Also Bought