Spécifications
- Length:317.5 mm (12.5")
- Description:Merit™ Simpson Uterine Sound
- Material:Silver Plated Stainless Steel
- Cat. no.:10185-322
- Supplier No.:98-382
Spécifications
A propos de cet article
The Merit™ Simpson Uterine Sound is an office-grade, malleable, silver plated OB/GYN instrument used for probing and dilating the uterus, as well as measuring the length of the cervical canal.
- It is round and thin with a small nodule at the tip and a thumb grip handle
- This product has a gently curved tip, graduated centimeter markings, and an overall length of 121/2"
It is very similar to the Sims uterine sound.