Double HEPA Filtration Kit, Labconco®
Supplier: Labconco
Certificates
About this item
Kit provides a second level of HEPA Filtration to meet United States Pharmacopeia (USP) <800> standard for redundant HEPA Filter in series.
Each kit includes one HEPA Filter (5’ and 6’ models include two), set of bag and straps for bag-in/bag-out system, complete metal support system, as well as certified installation instructions. The kit will add 32.7 cm (12.9") to the height of an existing system.
Kits are intended for field-installation atop XPert balance filtered systems and XPert bulk powder filtered systems (sold separately).
It features bag-in/bag-out HEPA filter(s) for protection during filter changes and durable steel construction.
Ordering information: Contact your Avantor sales representative for optional Canopy Connection for use with this kit.
Customers Who Bought This Also Bought