ContactUsLinkComponent
APO-BRDU™ kit, TUNEL Assay
APO-BRDU™ kit, TUNEL Assay
# de catalogue 10820-968
Fournisseur:  Tonbo Biosciences
Numéro CAS:  
APO-DIRECT* kit
APO-BRDU™ kit, TUNEL Assay
# de catalogue 10820-968
Fournisseur:  Tonbo Biosciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    1 Kit (50 sa mples)
  • Cat. no.:
    10820-968
Spécifications
A propos de cet article
The APO-DIRECT™ reagent kit provides a two color TUNEL (Terminal deoxynucleotide transferase dUTP Nick End Labeling) assay for labeling DNA breaks and total cellular DNA to detect apoptotic cells by flow cytometry. The kit contains the instructions and reagents required for measuring apoptosis in cells including: positive and negative control cells for assessing reagent performance; washing, reaction and rinsing buffers for processing individual steps in the assay; terminal deoxynucleotidyl tranferase enzyme (TdT), fluorescein isothiocyanate (FITC) deoxyuridine triphosphate (FITC-dUTP) for labeling DNA breaks and PI/RNase A solution for counter staining the total DNA.
  • Application: Flow Cytometry
Contents:
The APO-DIRECT Kit is shipped in one container that houses two packages. One package is shipped at ambient temperature and should be stored at 2-8° C upon arrival. The other package is in styrofoam, containing frozen ice packs. The contents of this styrofoam container should be stored at -20° C upon arrival. We have determined this shipping method is adequate to maintain the integrity of the kit components. Upon arrival store the reagents at the appropriate temperatures. Reagent bottles have color coded caps to aid in their identification. Sufficient reagents are provided to process 50 cell suspensions. Included are 5 mL positive and 5 mL negative control cell suspensions of approximately 1 x 106 cells per ml in 70% (v/v) ethanol. The control cells are derived from a human lymphoma cell line and have been fixed as described in the fixation procedure.
Delivery: The APO-DIRECT Kit is shipped in one container and consists of two packages. Upon arrival one should be stored at 2-8 °C and the other at -20 °C.
Customers Who Bought This Also Bought