Post Baskets and Mobile Basket Units, Quantum Storage Systems
Fournisseur: QUANTUM STORAGE SYSTEMS
Certificats
A propos de cet article
Industrial grade chrome wire with a 5/8" x 5/8" mesh design on the bottom/flat surface
Unique design allows baskets to be used without needing a flat shelves to stablize overall unit. Drop front and sloped sides provide super easy access, while providing a captive space for stored items.
Baskets are all adjustable on 1" increments utilizing standard 1" diameter posts. Casters included with mobile units.
Heavy-duty design provides a 600-800 lbs. load capacity when evenly distributed.
Customers Who Bought This Also Bought