Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Ext.Ø:82 mm
- Capacity:250 ml
- Height:59 mm
- Cat. no.:AA36098-KT
- Supplier No.:36098-KT
Spécifications
À propos de cet article
In the analytical laboratory, nickel crucibles offer high resistance to dilute alkalies at a very low cost per crucible.
- Life expectancy of a nickel crucible is from 4 to 6 fusions
- Straight wall design
- Completely resistant to phosphoric acid
- Highly resistant to the corrosive effect of the strongest alkalies
- Ideal for use in fusions where zirconium or other metals cannot be used
No dry gases are actively corrosive to nickel at atmospheric temperature. Nickel is also resistant to dry hydrogen chloride, hydrogen fluoride, and chlorine up to about 535°C. Nickel is not affected by steam at temperatures usually encountered. It is corroded by gases containing sulfur.