Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Length:127 mm (5")
- Material:Stainless steel
- Cat. no.:89025-550
- Supplier No.:15-3320
- Blade Type:Curved, Micro-Serrated, Blunt/Blunt
Spécifications
À propos de cet article
Sklarcut™ Ragnell Scissors, known as Kilner/Ragnell scissors are finger ring scissors, that feature micro-serrations on one blade to reduce trauma when cutting both delicate and course tissue.
These serrations assist in securely holding the tissue during cutting and minimizes tissue from slipping, so the most accurate and clean cuts can be made. The long shanks and short jaws allow the user greater control with less effort during dissection. This combined with flat blades and blunt tips often make Ragnell scissors the dissection instrument of choice during longer procedures.
Sklarcut™ scissors are identified by a single gold coated finger ring on the handle of the serrated blade.