Solvent Reservoirs, Plastic Coated, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Complete reservoir system includes a plastic-coated graduated bottle, a standard cap assembly, solvent and sparging filters.
- High performance
- Low pressure liquid chromatography mobile phase
Cap assembly fits GL45 threaded bottles and has a PTFE bulkhead with three ¹/₄"-28 threaded ports to accommodate fittings for ¹/₈" Ext.Ø tubing.
Customers Who Bought This Also Bought