Reagent Reservoirs, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
Reagent reservoirs provide a variety of solutions to suit all pipetting procedures.
- Graduations on inside wall enable quick measurement of remaining liquid
- Maximizes recovery if using only single channel and small amount of reagent
- Pour-off spouts and Extra-wide base adds rigidity and stability to reservoir, helping to avoid spills
Packaging: Sterile reservoirs packaged in snap-and-tear bags maintains sterility.
Customers Who Bought This Also Bought