Injector Lubricant for Agilent HPLC Systems, Restek
Catalog # 10057-992
Supplier: Restek
Specifications
- Description:Injector Lubricant, PTFE, Similar to Agilent# 79841-65501
- Size:10 g
- Cat. no.:10057-992
- Supplier No.:26520
Specifications
About this item
For use with Agilent 1050 and 1100 HPLC systems.
Customers Who Bought This Also Bought