Acid Neutralizing Kit, 6.5 Gallon, Brady
# de catalogue 10809-768
Fournisseur: Brady Worldwide
Spécifications
- Description:Acid Neutralizing Spill Kit
- Capacity:5 gal.
- Dimensions:18H x 14" Dia.
- Cat. no.:10809-768
- Supplier No.:SKA-BKTACID
Spécifications
A propos de cet article
Includes: 6.5 Gallon bucket (same as our SKA-BKT), 1 - 2lb shaker acid neutralizer, 10 Universal Pads, 2 Socs, 2 Bags, Goggles, Gloves, Tyvek Suit
Ordering information: (10) 15 in x 19 in Pads; (2) 3 in x 4 ft Socks; (2) Disposal Bags; 2 lb Acid Neutralizer Shaker; Goggles; Hazmat; Instruction Sheet; Nitrile Gloves; Scoop.
Customers Who Bought This Also Bought