ContactUsLinkComponent
Condenser Box
Condenser Box
# de catalogue 470219-540
Fournisseur:  Wards
Condenser Box
Condenser Box
# de catalogue 470219-540
Fournisseur:  Wards
Spécifications
  • Description:
    Condenser Box
  • Cat. no.:
    470219-540
  • Supplier No.:
    PH1136
Spécifications
A propos de cet article
This condenser box provides different values of Electrolytic capacitors, ranging from 1μF to 4.7μF. Built on a 8 x 5" plastic box. All these capacitors are of ± 10% tolerance and are of 63 volts working voltage.
Customers Who Bought This Also Bought