ContactUsLinkComponent
Rod Friction Set
Rod Friction Set
# de catalogue 470219-536
Fournisseur:  Wards
Electrostatic Friction Rod kit
Rod Friction Set
# de catalogue 470219-536
Fournisseur:  Wards
Spécifications
  • Description:
    Rod Friction Set
  • No. of Lab Groups:
    1
  • Cat. no.:
    470219-536
  • Supplier No.:
    PH1120150
Spécifications
A propos de cet article
Economical set for studying friction.
  • Exploration into electrostatic forces
This electrostatic kit provides the necessary tools to free both positive and negative charges for exploration into electrostatic forces. Students can experiment with different combinations of cloth and rod materials to explore how each manufactures charge carriers.
Instruction manual included.
Kit includes:
3 Fabric Cloths (wool, cotton, silk)
3 Rods (glass, ebonite, Lucite)
Customers Who Bought This Also Bought