ContactUsLinkComponent
Conductors; Set of Three
Conductors; Set of Three
# de catalogue 470219-522
Fournisseur:  Wards
Conductors
Conductors; Set of Three
# de catalogue 470219-522
Fournisseur:  Wards
Spécifications
  • Description:
    Conductors; Set of Three
  • Cat. no.:
    470219-522
  • Supplier No.:
    PH0880
Spécifications
A propos de cet article
Made of brass, interchangeable on pillars, fitted with insulators of ebonite on base. Spheres 5cm diameter, Cylinder 12.5 x 5cm diameter, Cone 10 x 5cm length x diameter.
Customers Who Bought This Also Bought