Quartz Wool, Velp Scientific
# de catalogue 10184-986
Fournisseur: VELP SCIENTIFIC INC.
Spécifications
- Description:Quartz Wool
- Size:50 g
- Cat. no.:10184-986
- Supplier No.:A00000154
Spécifications
A propos de cet article
Quartz wool for use with the DUMAS Nitrogen/Protein Analyzer.
Customers Who Bought This Also Bought