UVP Pen-Ray® Mercury Lamps, Analytik Jena
Supplier: Analytik Jena US
Certificates
About this item
Lamps are manufactured from the highest grade of quartz material.
These lamps are used for sterilization, fluorescent inspection, photochemical reaction, interferometry, chromatography, photometric and pluorometric instruments, spectroscopic wavelength calibration, monochromatic sources for optical systems and reference studies.
Customers Who Bought This Also Bought