Cleanroom Tape, Polyimide, UltraTape
Fournisseur: Ultratape Industries
Certificats
A propos de cet article
High temperature polyimide cleanroom tape with a silicone adhesive.
Customers Who Bought This Also Bought
High temperature polyimide cleanroom tape with a silicone adhesive.