DuPont™ ProShield® 10 Coveralls with Attached Hood and Boots
Fournisseur: DuPont
Certificats
A propos de cet article
DuPont™ ProShield® 10, spunbond meltblown spunbond (SMS) garments, are made to the high quality standards you have come to expect from DuPont.
- Standard fit hood
- Elastic wrists
- Attached skid-resistant boots
- Elastic waist
- Serged seams
ProShield® 10 garments have been optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments. DuPont™ ProShield® 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Applications include general maintenance, janitorial/cleaning and other dirty work. Attached hood with elastic around face opening is a standard fit hood. Elastic openings at wrists allows for a tighter fit. Attached boots feature skid-resistant soles.
Certifications: Thickness meets ASTM D1117.
Spécifications
- Catalog No:
- 10016-196
- 10016-218
- 10016-198
- 10016-220
- 10016-200
- 10016-202
- 10016-214
- 10016-194
- 10016-216
- 10016-190
- 10016-212
- 10016-192
- 10016-182
- 10016-204
- 10016-184
- 10016-206
- 10016-186
- 10016-208
- 10016-188
- 10016-210
- Garment application:
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
Spécifications
Informations connexes