DuPont™ ProShield® 10 Coveralls with Laydown Collar and Open Wrists
Supplier: DuPont
Certificates
About this item
DuPont™ ProShield® 10, spunbond meltblown spunbond (SMS) garments, are made to the high quality standards you have come to expect from DuPont.
- Laydown collar
- Open wrists and ankles
- Elastic waist
- Serged seams
ProShield® 10 garments have been optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments. DuPont™ ProShield® 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Applications include general maintenance, janitorial/cleaning and other dirty work. Coveralls feature a laydown collar and open wrists and ankles.
Certifications: Thickness meets ASTM D1117.
Specifications
- Catalog No:
- 10016-174
- 10016-154
- 10016-176
- 10016-156
- 10016-178
- 10016-158
- 10016-180
- 10016-168
- 10016-148
- 10016-170
- 10016-742
- 10016-150
- 10016-172
- 10016-744
- 10016-152
- 10016-746
- 10016-160
- 10016-162
- 10016-164
- 10016-166
- Garment application:
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
Specifications
Related Information
Product Information
Customers Who Bought This Also Bought