DuPont™ ProShield® 60 Boot Covers with PVC Sole
Fournisseur: DuPont
Certificats
A propos de cet article
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
- PVC skid-resistant soles
- Serged seams
- Elastic top
- Coverage up to calf
ProShield® 60 garments are made with a microporous film that is laminated to a spunbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation.
These 18" boot covers provide coverage up to the calf and feature an elastic top and PVC soles for enhanced skid-resistance and durability.
Certifications: Thickness meets ASTM D751.
Spécifications
- Catalog No:
- 10016-140
- Garment application:
- Laboratory