DuPont™ ProShield® 60 Coveralls with Respirator Hood
Supplier: DuPont
Certificates
About this item
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
- Respirator fit hood
- Elastic wrists and ankles
- Storm flap
- Serged seams
ProShield® 60 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation. Attached respirator-fit hood with elastic around face opening was designed to cover neck and chin and fit around respirator face mask. Longer zipper extends to chin for complete coverage of neck area. Storm flap of garment material covers zipper closure. Elastic openings at wrists and ankles allows for a tighter fit.
Certifications: Thickness meets ASTM D751.
Specifications
- Catalog No:
- 10016-110
- 10016-112
- 10016-114
- 10016-124
- 10016-116
- 10016-118
- 10016-120
- 10016-122
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
Specifications
Related Information
Product Information
Customers Who Bought This Also Bought