QualaSheer® VHC12 QualaSheer XC Cleanroom Vinyl 12" Gloves, QRP
Fournisseur: PIP
Certificats
A propos de cet article
The QRP VHC12 ESD-Safe Clean Room Class 100 Vinyl ISO 5 Gloves are 100% PVC co-polymer (no latex) for ISO 5 (Class 100) applications.
- Naturally anti-static; 10¹⁰ ohms
- Microtextured
Certifications: Meets or exceeds all applicable requirements of IEST-RP-CC005.2.
Spécifications
- Catalog No:
- 89040-962
- 89040-956
- 89040-958
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5