A propos de cet article
Ranger® 4000 Compact Industrial Scales offer the best value for durable industrial weighing applications. These units feature the largest, white LED display on any scale in its class. Combined with checkweighing LEDs and audible indicators, this makes the Ranger® 4000 highly functional in loud and dim industrial settings.
- 100-hours continuous use (12-hour recharge time)
- Internal rechargeable lead acid battery
All models feature a stainless steel platform, integral weigh-below hook, sealed front panel, menu lock switch, up-front level indicator, adjustable leveling feet, selectable environmental and auto-print settings, stability indicator, overload and underload indicators, low battery indicator, auto shut-off, and auto tare.
Additional standard features include; die cast aluminum housing, a variety of weighing units (kg, g, lb, oz, lb:oz), a variety of application modes (Weighing, Percent Weighing, Parts Counting, Checkweighing, Checkcounting, Checkpercent, Accumulation, Dynamic Weighing/Display Hold), and a 40mm (1.5") white LED display with 6-digit, 7-segment capabilities.
Spécifications
- Catalog No:
- 10820-780
- 10820-782
- 10820-776
- 10820-778
- Operating temperature range:
- –10 to 40°C (14 to 104°F)
- –10 to 40°C (14 to 104°F)
- –10 to 40°C (14 to 104°F)
- –10 to 40°C (14 to 104°F)
- Communication interface:
- Built-in RS-232, Optional 2nd RS-232, USB, Ethernet
- Built-in RS-232, Optional 2nd RS-232, USB, Ethernet
- Built-in RS-232, Optional 2nd RS-232, USB, Ethernet
- Built-in RS-232, Optional 2nd RS-232, USB, Ethernet
- Voltage:
- 100–240V, 50/60Hz (Internal Universal Power Supply)
- 100–240V, 50/60Hz (Internal Universal Power Supply)
- 100–240V, 50/60Hz (Internal Universal Power Supply)
- 100–240V, 50/60Hz (Internal Universal Power Supply)