About this item
This pH buffer kit has colour coded buffers, storage solution and storage bottles.
They are manufactured under ISO 9000 quality standards. The kit includes 1 pint (475 ml) bottle each of pH 4.00 buffer, pH 7.00 buffer, pH 10.01 buffer and standard pH electrode storage solution and one pH electrode storage bottle.
pH buffers with temperature tables on the labels are avilable for accurate calibrations and NIST™ traceability.