Sealy Dissecting Scissors, OR Grade, Sklar
Fournisseur: Sklar
Certificats
A propos de cet article
Sklar's Sealy Dissecting Scissors are finger ring scissors used for dissecting tissue.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.