ContactUsLinkComponent
Vasectomy Piercing Forceps, OR Grade, Sklar
Vasectomy Piercing Forceps, OR Grade, Sklar
# de catalogue 10192-390
Fournisseur:  Sklar
Forceps
Vasectomy Piercing Forceps, OR Grade, Sklar
# de catalogue 10192-390
Fournisseur:  Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Length:
    140 mm (5 1/2")
  • Material:
    Stainless Steel
  • Tip Configuration:
    Sharp
  • Cat. no.:
    10192-390
  • Tip Style:
    Curved
  • Supplier No.:
    85-6224
Spécifications
A propos de cet article
Vasectomy Piercing Forceps are a ratcheted, finger ring instrument with sharp tips used to pierce the scrotal skin in sutureless vasectomy procedures
It is used in conjunction with the vasectomy clamp with ring tips.
Customers Who Bought This Also Bought