Spécifications
- Length:140 mm (5 1/2")
- Material:Stainless Steel
- Tip Configuration:Sharp
- Cat. no.:10192-390
- Tip Style:Curved
- Supplier No.:85-6224
Spécifications
A propos de cet article
Vasectomy Piercing Forceps are a ratcheted, finger ring instrument with sharp tips used to pierce the scrotal skin in sutureless vasectomy procedures
It is used in conjunction with the vasectomy clamp with ring tips.