Hydrion® Single Roll Dispenser, pH 1.0 to 12.0, Micro Essential Laboratory®
Supplier: Micro Essential
Certificates
About this item
Hydrion® Insta-Chek pH paper is an excellent direct reading pH paper with the widest range available in a single paper.
- Simple and single color match
- Distinct and easily matched color separations
- Accurate, reproducible readings with an immediate response
General purpose pH papers has a distinct color match at full pH units.
Packaging: Each carton contains 10 kits, each consisting of a 15-foot roll of test paper and matching color chart. Provides for approximately 1000 tests.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.