VWR® Serological Pipette
# de catalogue 53283-490
Fournisseur: VWR International
Spécifications
- Capacity:5 ml
- Cat. no.:53283-490
- Tolerance:±0.10 ml
- Subdivisions:0.1 ml
- Negative Graduations:2 ml
Spécifications
A propos de cet article
These serological plastic pipettes are designed to use with tissue culture work.
- Black printed scale
- Calibrated to deliver to the tip, with two rings near the top
- Irradiation sterilized
Customers Who Bought This Also Bought