About this item
Sklarlite™ Mayo Dissecting Scissors are a lighter, more delicate version of the standard Mayo scissors
While the curved instrument is used for cutting or dissecting dense or deep tissue, and the straight version is used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference.
Sklar also offers Mayo Dissecting Scissors with a selection of special features to meet procedure specifications and surgeon preferences.
Sklar's lightweight modification of this versatile instrument that can be used in multiple procedures and settings provides the surgeon with greater control and less fatigue.