Dropping Bottles, Glass, G.S.C. INTERNATIONAL, INC.
Fournisseur: GSC International
Certificats
A propos de cet article
Heavy flint glass.
- Come with glass pipet and rubber bulb in screw cap
- Use amber bottles for light-sensitive liquids
Customers Who Bought This Also Bought