Spécifications
- Description:Sklar Graves Wide View Vaginal Speculum, Medium, Straight
- Material:Stainless Steel
- Cat. no.:10197-208
- Supplier No.:90-3705
Spécifications
A propos de cet article
Graves wide view vaginal speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix.
It is a standard double blade duckbill speculum that features a wide proximal opening for instrumentation. It is inserted with the blades closed. The blades are then opened by an external lever and screw.