Pierce™ Protein A Spin Plate for IgG Screening, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Pierce™ Protein A and Protein G Spin Plates for IgG Screening offers a high throughput format for quick purification and screening of antibodies and for immunoprecipitation (IP)
- Immobilized Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum
- Agarose resin
- Inert and stable
- High capacity
The Protein A and Protein G resin purifies by binding IgGs, primarily through their Fc region, in a buffer that facilitates the interaction. Non-relevant proteins, such as albumin and transferrin, are removed by washing and the purified antibody is then recovered using low-pH elution conditions.
Each well of the 96-well spin plate can process 10 to 100 µl of serum, cell culture supernatant or ascites fluid samples.
Delivery: Each pack contains 1 plate (each well contains 50 µl of resin for processing 10 to 100 µl of sample) and 3 wash/collection plates.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Spécifications
- Catalog No:
- CAPI45204
- CAPI45202
- Storage temperature:
- Upon receipt store at 4˚C.
- Upon receipt store at 4˚C.