SGPF Series Sterile Powder Free Latex Gloves, CT International
Fournisseur: CT International
Certificats
A propos de cet article
SGPF Series are sterile powder free gloves. They are manufactured for outstanding strength, stretch and fit.
- Textured surface and hand specific
- Low protein, double chlorinated and contact sensitizers
- Tapered, beaded cuff
- Folded cuff for aseptic donning
- Gamma irradiated sterilization minimum dose 2.5 mrads
They provides a textured surface and are double chlorinated reducing contact sensitizers and protein content. SGPF Series gloves are lot traceable.
Spécifications
- Catalog No:
- 56617-168
- 56617-170
- 56617-172
- 56617-174
- 56617-176
- 56617-178
- 56617-180
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- FDA regulated:
- +
- +
- +
- +
- +
- +
- +
- ASTM No.:
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05