PFXT Series Class 100 Powder Free Gloves, CT International
Supplier: CT International
Certificates
About this item
PFXT series are class 100 powder free gloves.
- Manufactured for outstanding strength, stretch and fit
Offer a textured surface and are double chlorinated reducing contact sensitizers and protein content.
Certifications: PFXT Series gloves are ISO 5 Clean Room compatible or higher for controlled environments.
Specifications
- Catalog No:
- 56617-246
- 56617-232
- 56617-234
- 56617-236
- 56617-238
- 56617-240
- 56617-242
- 56617-244
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- ASTM No.:
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05