CHS Series Latex Water Washed Class 10 Gloves, CT International
Fournisseur: CT International
Certificats
A propos de cet article
CHS Series are latex DI water washed Class 10 gloves.
- Offers textured surface
They double chlorinated reducing contact sensitizers and protein content.
Like all our gloves, they are manufactured for outstanding strength, stretch and fit.
Certifications: CHS Series Class 10 Latex gloves ISO 4 Clean Room compatible or higher for controlled environments.
Spécifications
- Catalog No:
- 56613-806
- 56613-740
- 56613-741
- 56613-742
- 56613-746
- 56613-692
- 56613-694
- 56613-739
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- ASTM No.:
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05
- D3578-05