NCP Series Nitrile Co-Polymer Powder Free Class 100 Gloves, CT International
Fournisseur: CT International
Certificats
A propos de cet article
NCP Series gloves are class 100 gloves are manufactured for outstanding strength, stretch and fit.
- Offer a textured surface and a super soft feel for comfort
- Nitrile co-polymer gloves
- Powder free
Certifications: These gloves are ISO 5 clean room compatible or higher controlled environments.
Spécifications
- Catalog No:
- 56613-858
- 56613-860
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- ASTM No.:
- D3578-95
- D3578-95