Large Lab Type Scalpel Handle, OR Grade, Sklar®
Supplier: Sklar
Certificates
About this item
Sklar's Large Lab Type Scalpel Handles are designed for use in sharp dissection
These handles are phenolic versus stainless steel, and thicker and shorter than the stainless steel handles.
Ordering information: Each sold separately.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.