BioHazardous Waste Cans, Eagle Mfg
Fournisseur: Eagle Manufacturing
Certificats
A propos de cet article
These waste cans are made of red high-density polyethylene (HDPE).
- Meets OSHA requirements for exposures to bloodborne pathogens
- Red color with biohazard symbol
Certifications: OSHA 29 CFR 1910.1030 and FM approved.
Customers Who Bought This Also Bought