Admittance and Security Signs without Header, National Marker
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
Alert visitors and guests of security policies and direct them towards appropriate entrances.
- Available in alum, PS vinyl, rigid plastic (thick polystyrene), glow polyester, and glow rigid plastic materials
- 0.040 alum is weatherproof, non-corrosive, and has baked-on enamel on both sides
- 0.004 PS vinyl material has pressure sensitive adhesive backing
- 0.050 rigid plastic material has radius corners and pre-drilled corner mounting holes
- 0.010 glow polyester film and 0.050 glow rigid plastic materials are non-toxic, non-radioactive, and glow for more than 6 hours
Customers Who Bought This Also Bought