PIPETBOY acu 2 Pipette Aid, INTEGRA Biosciences
Fournisseur: INTEGRA Biosciences
Certificats
A propos de cet article
This second generation model, the PIPETBOY acu 2 Pipette Aid, is equipped with the latest technology to provide increased pipetting speed and a significant weight reduction in the instrument.
These two features help users to cut work time while offering an improved ergonomic pipetting experience. Besides these updates, the main benefits of the PIPETBOY acu Pipette Aid were preserved, including the unmatched pipetting precision, the colorful look, and the weight balance. Lighter and faster, the world’s most popular pipettor just got better.
Ordering information: PIPETBOY acu 2 Pipette Aids are supplied complete with Li-Ion battery, mains adapter, spare sterile 0.45 µm filter, and wall mount. A free Swiss pocket tool is included with each new PIPETBOY acu 2.
Spécifications
- Catalog No:
- 14222-180
- 37001-862
- 37001-860
- 37001-858
- 37001-856
- 37001-864
- Max. speed:
- 13.5 ml/sec. (with a 50 ml pipet)
- 13.5 ml/sec. (with a 50 ml pipet)
- 13.5 ml/sec. (with a 50 ml pipet)
- 13.5 ml/sec. (with a 50 ml pipet)
- 13.5 ml/sec. (with a 50 ml pipet)
- 13.5 ml/sec. (with a 50 ml pipet)
- Power:
- Input: 100–240 VAC, 50/60 Hz, Output: 17 VDC ±10 %/180 mA ±10 %
- Input: 100–240 VAC, 50/60 Hz, Output: 17 VDC ±10 %/180 mA ±10 %
- Input: 100–240 VAC, 50/60 Hz, Output: 17 VDC ±10 %/180 mA ±10 %
- Input: 100–240 VAC, 50/60 Hz, Output: 17 VDC ±10 %/180 mA ±10 %
- Input: 100–240 VAC, 50/60 Hz, Output: 17 VDC ±10 %/180 mA ±10 %
- Input: 100–240 VAC, 50/60 Hz, Output: 17 VDC ±10 %/180 mA ±10 %
- Battery type:
- Li-Ion, min. 500 mAh
- Li-Ion, min. 500 mAh
- Li-Ion, min. 500 mAh
- Li-Ion, min. 500 mAh
- Li-Ion, min. 500 mAh
- Li-Ion, min. 500 mAh
- Weight:
- 195 g (0.43 lbs.)
- 195 g (0.43 lbs.)
- 195 g (0.43 lbs.)
- 195 g (0.43 lbs.)
- 195 g (0.43 lbs.)
- 195 g (0.43 lbs.)