ContactUsLinkComponent
Calbiochem®, Phosphatase Inhibitor Cocktail Set II, EMD Millipore
Calbiochem®, Phosphatase Inhibitor Cocktail Set II, EMD Millipore
# de catalogue CA80501-130
Fournisseur:  MilliporeSigma
Numéro CAS:  
Calbiochem®, Phosphatase Inhibitor Cocktail Set II, EMD Millipore
# de catalogue CA80501-130
Fournisseur:  MilliporeSigma
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Phosphatase inhibitors
  • Size:
    1 set (5 x 1 mL)
  • Cat. no.:
    CA80501-130
  • Supplier No.:
    524625-1SET
Spécifications
A propos de cet article
A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs)
Suitable for use with cell lysates and tissue extracts, including samples containing detergents.
Customers Who Bought This Also Bought