Sintered Glass Fiber Filter Discs, Porosity B, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These glass fiber filter discs last longer and are more abrasion resistant.
- Superior in performance to granular type discs
- Resistance to thermal shock and chemical attack
- Fabricated from borosilicate glass
Available in a complete range of porosities and diameters, these discs can be used in the manufacture of a wide variety of special apparatus.
Customers Who Bought This Also Bought