Caution Warehouse Signs, National Marker
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
Safety signs are the perfect way to alert your employees and guests when a hazard is present in your facility
- Ideal for indoor and outdoor labeling
- Available in Alum, PS vinyl and Rigid plastic (thick polystyrene) materials
- .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
- .004 PS vinyl material has pressure sensitive adhesive backing
- .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
These signs point out easily overlooked dangers in and around the workplace. Avoid accidents and keep employees safe by clearly identifying hazardous equipment situations. This can include heavy equipment, moving machine parts and powered vehicles.
Customers Who Bought This Also Bought