A propos de cet article
Contains formula N (tetrafluoroethane).
- 100% safe for the ozone
- Contains no flammable elements
It is provided with a patented 360° vector valve. The multi-directional valve is used for easy blasting in hard to reach areas. The valve can be removed from the can anytime without any loss of gas.