Personal Protection (PPE), OSHA Caution Signs, National Marker
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
OSHA signs are the perfect way to alert employees and guests when a hazard is present in the facility.
- Ideal for indoor and outdoor labeling
- Available in aluminum, PS vinyl, and rigid plastic (thick polystyrene) materials
- 0.040 aluminum is weatherproof, non-corrosive and has baked-on enamel on both sides
- 0.004 PS vinyl material has pressure sensitive adhesive backing
- 0.050 rigid plastic material has radius corners and pre-drilled corner mounting holes
Keep facilities safe by displaying an OSHA regulated sign today, and prevent an accident tomorrow. Each sign header reads, "Caution."
Certifications: Comply with OSHA 29 CFR 1910.
Customers Who Bought This Also Bought