Crimpers and Decappers for Microwave Vials, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Tools for crimping and decapping Aluminum seals on Microwave vials.
The crimper has an adjustment feature which will allow it to be used with a variety of septa, discs, stoppers or lined caps. The user may adjust the stroke or movement for each application as needed. The pliers decapper is an inexpensive way to remove seal caps by simply securing the seal between the jaws and squeezing the handles until cap pops off.
Customers Who Bought This Also Bought