To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Clonality:Polyclonal
- Gene ID:ACTL6A/86; ACTL6B/51412
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:BAF53A + BAF53B
- Conjugation:Unconjugated
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:ACTL6|INO80 complex subunit K|hArpN alpha|BAF53A|BAF complex 53 kDa subunit|BRG1-associated factor|actin-like 6B|BAF53b|BAF53|53 kDa BRG1-associated factor B|actin-related protein 4|ACTL6A|MGC5382 Arp4|actin-like 6A|BAF53B|ARPN-BETA|actin-related protein|BAF53a|INO80K|hArpN beta|ACTL6B
- Storage buffer:TBS, pH 7.3, with 0.5% BSA and 0.02% Sodium azide
- Storage temperature:–20 °C, avoid freeze/thaw cycles
- Cat. no.:CAPIPA5-18272
- Supplier No.:PA5-18272
Spécifications
A propos de cet article
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. The protein encoded by this gene is a member of a family of actin-related proteins which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene encodes a subunit of the BAF complex in mammals, which is functionally related to SWI/SNF complex in S. cerevisiae and Drosophila; the latter is thought to facilitate transcriptional activation of specific genes by antagonizing chromatin-mediated transcriptional repression. This subunit may be involved in the regulation of genes by structural modulation of their chromatin, specifically in the brain.
Western Blot: 1-3 ug/ml
Type: Primary
Antigen: BAF53A/BAF53B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human