Acetate Buffer Solution, pH 4.0, VWR Chemicals BDH®
Supplier: VWR International
Certificates
About this item
APHA 4500-C1 and ASTM 1253-91 for chlorine.
Standardized at 25 °C against chemicals whose certification is traceable to NIST.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.