Spécifications
- Description:Sklar Black™ Auvard Weighted Vaginal Speculum
- Material:Stainless Steel with Reduced Reflectivity Polymer Coating
- Tip Configuration:Straight, Smooth, Blunt
- Cat. no.:10184-284
- Supplier No.:90-9108
Spécifications
A propos de cet article
Sklar Black™ Auvard Weighted Vaginal Speculum is a single-end vaginal speculum commonly used in laser OB/GYN procedures
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
It is available with various blade styles and three weight choices in order to accommodate the needs of the patient and the procedure. The weight holds the speculum in place and provides an open view for the surgeon to perform procedures